Water, purified, sterile

Supplier: VWR Chemicals

VWRCAX021376EA 281 SGD
VWRCAX021376
Water, purified, sterile
Water
Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Aspect Limpid
Volume (ml) 90 ± 1,8 ml
Membrane filtration according to PE 2.6.1 Sterile after 14 days (Incubation 20-25°C & 30-35°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR