VWR®, Aprotinin
Supplier: VWR Chemicals
VWRCE429-50MGEA 222 SGD
VWRCE429-50MG
VWR®, Aprotinin
Proteins and Peptides
A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin. Not an inhibitor of Factor Xa and thrombin.
Activity (KIU/mg)      4500