Water for biotechnology nuclease-free, 0.2 µm filtered

Supplier: VWR Chemicals

Synonyms: Hydrogen oxide

Filtered through a 0.2 µm filter.

VWRCE476-500MLEA 74.5 SGD
VWRCE476-500ML VWRCE476-1L VWRCE476-100ML
Water for biotechnology nuclease-free, 0.2 µm filtered
Water

Nuclease-free water treated with diethylpyrocarbonate (DEPC) for nucleic acid applications.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

DNase NONE
Protease NONE
RNase NONE
Sterility PASS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR