Hydrochloric acid 6 mol/l (6 N), Reagent Grade

Supplier: VWR Chemicals
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

VWRCE484-500MLEA 155 SGD
VWRCE484-500ML VWRCE484-1L
Hydrochloric acid 6 mol/l (6 N), Reagent Grade
Hydrochloric acid
Formula: HCl
Density: 1,06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Concentration 5.4 – 6.6 moles/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR