VWR®, Phosphate buffered saline (PBS) 1X, for biotechnology
Supplier: VWR Chemicals
VWRCE504-500MLEA 51.5 SGD
VWRCE504-500ML
VWR®, Phosphate buffered saline (PBS) 1X, for biotechnology
Buffers Cell Culture Buffers
137 mM sodium chloride, 2,7 mM potassium chloride, 12 mM phosphate buffer
  • pH 7,4
Order Now
SPECIFICATIONS
Environmentally Preferable