Dextran sulphate sodium salt, M.W. ~ 500,000 50% for biotechnology

Supplier: VWR Chemicals
Warning

Synonyms: Dextran sulphate sodium from Dextran

VWRCE516-100MLEA 506 SGD
VWRCE516-100ML
Dextran sulphate sodium salt, M.W. ~ 500,000 50% for biotechnology
Dextran sulphate sodium salt

Used for the detection of DNA or RNA in nucleic acid hybridisation procedures. It effectively “excludes” probe from solution, accelerating the rate of annealing.

Formula: (C₆H₇Na₃O₁₄S₃)n
MDL Number: MFCD00081551
CAS Number: 9011-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae52589f20d0da38bdceca63c873f7fba4580dd4&vid=bf56737a35089faf2295e8fcb2d8a1ea654b245a&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddbddce328004684d12a8dc9f1b65d847c433847&vid=f9e15aedab27c83fb2129e9d24b773e8c42b2a7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

APHA (1:10) Tan to yellow haze-free liquid
DNase none detected
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR