VWR®, MOPS 10X buffer for biotechnology
Supplier: VWR Chemicals
VWRCE526-500MLEA 431 SGD
VWRCE526-500ML
VWR®, MOPS 10X buffer for biotechnology
Buffers Electrophoresis Buffers Running Buffers
MOPS is a zwitterionic buffer used as a running buffer for denaturing agarose gel electrophoresis of RNA. Having a pH buffering range from 6,5 to 7,9, MOPS works exceptionally well with formaldehyde gels at 20 mM concentration.
  • Optimised for RNA agarose electrophoresis
  • Ideal for use with RNA formaldehyde gels
Order Now
SPECIFICATIONS
Environmentally Preferable