Sodium chloride 5 mol/l (5 N) in aqueous solution for biotechnology

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRCE529-500MLEA 119 SGD
VWRCE529-500ML
Sodium chloride 5 mol/l (5 N) in aqueous solution for biotechnology
Sodium chloride

Common source of salts for media propagation of many organisms.

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Conductivity (1 %, Water) 5000 - 7000 umhos
Identification PASS
pH @25 °C 5.0 - 9.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR