Orthophosphoric acid ≥85% ACS

Supplier: VWR Chemicals
Danger

Synonyms: Phosphoric acid

VWRCE582-500MLEA 162 SGD
VWRCE582-500ML VWRCE582-50ML
Orthophosphoric acid ≥85% ACS
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Antimony <0.002%
APHA ≤10
Arsenic ≤0.0001%
Assay ≤85%
Calcium ≤0.002%
Chloride ≤0.0003%
Heavy Metals ≤0.001%
Insoluble Matter ≤0.001%
Iron ≤0.003%
Magnesium ≤0.002%
Manganese ≤0.5 ppm
Nitrate ≤0.0005%
Potassium ≤0.005%
Reducing Substances Pass
Sodium ≤0.025%
Sulphate ≤0.003%
Volatile Acids ≤0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR