VWR®, Destaining bags
Supplier: VWR Chemicals
VWRCE732-25EA 256 SGD
VWRCE732-25
VWR®, Destaining bags
Electrophoresis Reagents Stain Removal Reagents
Destaining bags are designed to safely remove ethidium bromide from solution during overnight treatment. Each bag extracts up to 5 mg of ethidium bromide from solution, thus minimising the exposure of research personnel to this toxic material. The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration.
  • Removes up to 5 mg ethidium bromide per bag
  • Minimises exposure to toxic materials
  • Easiest ethidium bromide removal available