Cobalt(II) chloride hexahydrate 98-102% ACS

Supplier: VWR Chemicals
Danger

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

VWRCJ297-500GEA 229 SGD
VWRCJ297-500G VWRCJ297-100G
Cobalt(II) chloride hexahydrate 98-102% ACS
Cobalt (II) chloride hexahydrate
Formula: CoCl₂·6H₂O
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: 56 °C
Density: 1,92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 13,02462

Order Now

Specification Test Results

Assay 98 – 102%
Calcium ≤0.005%
Copper ≤0.002%
Insolubles ≤0.01%
Iron ≤0.005%
Magnesium ≤0.005%
Nickel ≤0.1%
Nitrate ≤0.01%
Potassium ≤0.01%
Sodium ≤0.05%
Sulphate ≤0.01%
Zinc ≤0.03%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR