VWR®, Yeast nitrogen base (YNB)
Supplier: VWR Chemicals
VWRCJ386-100GEA 578 SGD
VWRCJ386-100G VWRCJ386-500G
VWR®, Yeast nitrogen base (YNB)
Media for Microbiology Base Materials for Microbiology
Yeast Nitrogen Base (YNB) is a base medium for preparation of minimal and synthetic defined yeast media.
  • Animal origin
  • Suitable for studying strains of yeast requiring certain vitamins
Ordering information: Base materials are available with or without ammonium sulfate for the classification of yeast.