VWR®, Pepstatin A

Supplier: VWR Chemicals

VWRCJ583-25MGEA 762 SGD
VWRCJ583-25MG
VWR®, Pepstatin A
Proteins and Peptides

An aspartic acid protease inhibitor of pepsin, renin, and cathepsin D. Also an inhibitor of the HIV protease. Not water soluble.

Order Now

SPECIFICATIONS

Protein/peptide name Pepstatin A
Environmentally Preferable
Species
Source
CAS No. 26305-03-3
Grade Ultra Pure Grade
Molecular formula C₃₄H₆₃N₅O₉

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR