VWR® LB, Broth, Liquid, for Biotechnology
Supplier: VWR Chemicals
VWRCJ833-1LEA 131 SGD
VWRCJ833-1L
VWR® LB, Broth, Liquid, for Biotechnology
Media for Microbiology Prepared Media for Microbiology
LB Broth, liquid is used for the propagation and maintenance of E. coli. Widely used for the preparation of plasmid DNA and recombinant proteins.
  • Ready to use liquid media
  • Clear, yellow to brown liquid
One L of media contains 10 g sodium chloride, 10 g tryptone/case in peptone, 5 g yeast extract, bacteriological.
No growth detected in bioburden (Milliflex) (EQC) test.
pH (25 °C): 6,65 to 7,15.