VWR® 2XYT Medium Broth for Biotechnology
Supplier: VWR Chemicals
VWRCJ902-500GEA 461 SGD
VWRCJ902-500G
VWR® 2XYT Medium Broth for Biotechnology
Media for Microbiology Dehydrated Media for Microbiology
2XYT medium broth powder is used to prepare rich broth for the propagation of M13 bacteriophage.
  • Save time by using pre-mixed, ready to use media
  • Homogenous, high quality powders
  • Light beige, free flowing powder
31 g of powder when dissolved per litre of deionised water will yield 16 g/L of casein peptone, 10 g/L of casamino acids and five g/L of sodium chloride.
Order Now
SPECIFICATIONS
Environmentally Preferable