VWR® YPD, Broth, for Biotechnology
Supplier: VWR Chemicals
VWRCJ903-500GEA 516 SGD
VWRCJ903-500G
VWR® YPD, Broth, for Biotechnology
Media for Microbiology Dehydrated Media for Microbiology
Yeast peptide dextrose (YPD) broth is used for the propagation and maintenance of yeasts.
YPD broth is supplied as a pre-mixed powder which, when dissolved at 50 g/l of water, yields peptone (20 g/l), yeast extract (10 g/l), glucose (20 g/l).
Final pH of solution (25 °C): 6,5 ±0.5
Autoclave or sterile filter.
Order Now
SPECIFICATIONS
Environmentally Preferable