VWR®, DNA ladder, 100 bp
Supplier: VWR Chemicals
VWRCK180-250ULEA 197 SGD
VWRCK180-250UL
VWR®, DNA ladder, 100 bp
Electrophoresis Reagents Molecular Weight Markers
Ladder with 13 fragments ranging from 100 to 3000 bp for easy band identification.
Also available pre-mixed with loading dye.
Order Now
SPECIFICATIONS
Environmentally Preferable