VWR® BANDit™ Protein Stain Blue
Supplier: VWR Chemicals
VWRCK217-1LEA 342 SGD
VWRCK217-1L
VWR® BANDit™ Protein Stain Blue
Stains Electrophoresis Stains
Blue BANDit is a safe and environmentally friendly alternative to traditional Coomassie® Blue staining.
  • Reduces handling of hazardous chemicals: No methanol or acetic acid required
  • High sensitivity, quick, simple procedure