Methyl cellulose, 4000 mPa.S (2% H₂O), high purity

Supplier: VWR Chemicals

Synonyms: Cellulose methyl ether

VWRCK390-250GEA 721 SGD
VWRCK390-250G
Methyl cellulose, 4000 mPa.S (2% H₂O), high purity
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Specification Test Results

Solubility (2%, Water) Pass
Viscosity (2%, water) @20 °C 3500 – 5600 cps
Appearance white powder with a yellow cast


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR