Lithium chloride 8 mol/l aqueous solution Biotechnology

Supplier: VWR Chemicals
Warning

VWRCK445-100MLEA 409 SGD
VWRCK445-100ML
Lithium chloride 8 mol/l aqueous solution Biotechnology
Lithium chloride

A convenient, ready-to-use solution that helps to remove inhibitors of cDNA synthesis and translation.


  • High concentration for easy dilution
  • Nuclease and protease free
  • Does not precipitate DNA, protein, or carbohydrates


This Lithium Chloride Solution comes especially formulated for RT PCR preparation and cDNA synthesis. This solution is a nuclease-free, protease-free, 8 Molar preparation of lithium chloride in deionised water and is highly concentrated for easy dilution.


It is ideal for the precipitation of RNA right after total RNA isolation or in vitro transcription. The lithium chloride is the key to this process, as it efficiently precipitates the RNA, but does not precipitate the DNA, protein, or free nucleotide triphosphates.


To ensure a quality product throughout, this 8 M Lithium Chloride Solution is 0.2 micron filtered and packaged in a PETG Nalge bottle. Solution is nuclease and protease free, and it is ready-to-use.

Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

DNase None Detected
Protease None Detected
RNase None Detected
Titration 7.7 – 8.2 M
Appearance Clear, colorless liquid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR