VWR®, Bovine serum albumin (BSA)
Supplier: VWR Chemicals
VWRCK719-120MLEA 930 SGD
VWRCK719-120ML
VWR®, Bovine serum albumin (BSA)
Proteins and Peptides
Concentrated 30% BSA solution, fraction V, cold alcohol isolation, in 0,85% NaCl, contains 0,1% Na-azide (as a stabiliser).