VWR® WIPA-BOOTS Microorganism Pathogen Sampling in Livestock Industry
Supplier: VWR Chemicals
VWRCKSS-67410-TSEA 1580 SGD
VWRCKSS-67410-TS
VWR® WIPA-BOOTS Microorganism Pathogen Sampling in Livestock Industry
Bags Sample Bags
This product is a quick and simple solution to collect microobiological samples in livestock housing, animal transportation vehicles, and their washing stations. The purpose is the prevention, detection, and control of infections by viruses (H1N1, African swine fever, PRRS, Circovirus, Newcastle) and bacteria (Salmonella and E. coli including STEC).
  • Ideal for sample collection and inspection of surfaces in contact with livestock
  • Easy to use and highly versatile
  • Simultaneous sample collection and inspection
  • WIPA-BOOTS are impregnated with a solution that
More Product Information