Aprotinin (Trypsin inhibitor, pancreatic basic) (from bovine lung), proteomics grade

Supplier: VWR Chemicals

VWRCM167-10MGEA 340 SGD
VWRCM167-10MG
Aprotinin (Trypsin inhibitor, pancreatic basic) (from bovine lung), proteomics grade
Proteins and Peptides

A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin. Not an inhibitor of Factor Xa and thrombin.


Activity (KIU/mg)      ≤4500


Purity (HPLC)      Pass

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR