VWR®, Pepstatin A

Supplier: VWR Chemicals

VWRCM183-5MGEA 601 SGD
VWRCM183-5MG
VWR®, Pepstatin A
Proteins and Peptides

An aspartic protease inhibitor of pepsin, renin, and cathepsin D. Can also block HIV protease. Not soluble in water.


FTIR: Suitable
Melting point: 220-240 ° C
Optical rotation (0.5%, MeOH): -95 to -85 °
Purity (HPLC): 75%

Order Now

SPECIFICATIONS

Protein/peptide name Pepstatin A
Environmentally Preferable
Species
Source
CAS No. 26305-03-3
MDL No. MFCD00060740
Grade Proteomics Grade
Molecular formula C₃₄H₆₃N₅O₉
Storage conditions Refrigerator

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR