VWR® Protease Inhibitor Cocktail, Plant
Supplier: VWR Chemicals
VWRCM307-1MLEA 280 SGD
VWRCM307-1ML
VWR® Protease Inhibitor Cocktail, Plant
Cell Culture Supplements and Additives
Contains a mix of protease inhibitors: AEBSF, E-64, Bestatin, Pepstatin, Leupeptin and 1, 10-Phenanthroline. Each vial can be reconstituted in 1 ml dimethyl sulfoxide to form a 100X solution.