VWR®, Taq DNA polymerase, 500 U
Supplier: VWR Chemicals
VWRCN224-500UEA 447 SGD
VWRCN224-500U
VWR®, Taq DNA polymerase, 500 U
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Taq DNA Polymerase is ideal for general PCR and primer extension reactions to amplify DNA templates with high sensitivity. PCR can be initiated from as few as 6 primed templates. Simply add 0,5 µl of Taq DNA Polymerase to a 25 µl PCR reaction.
  • Highly purified, DNA-free, and free of bacterial genomic DNA
  • Amplify DNA templates with high sensitivity
  • Amplify single-copy and low abundance templates
One unit of activity incorporates 10 nmoles of dNTP into acid-insoluble DNA in 30 minutes at 72 °C.
The PCR kit with Taq DNA Polymerase includes separate solutions of all material needed for a PCR reaction, including dNTPs, reaction buffer with and without MgCl2, 25 mM MgCl2, Taq Polymerase, and sterile water.
Delivery information: Contains Taq DNA Polymerase, 500 U (5 U/µl) and Taq Buffer, 10X, 1 ml. Sufficient for 200× 25 µl amplifications.