D(-)-Luciferin sodium salt, Ultra Pure Grade

Supplier: VWR Chemicals

Synonyms: Sodium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate, D-Luciferin, sodium, D-Luciferin sodium, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt

VWRCN456-50MGEA 226 SGD
VWRCN456-50MG
D(-)-Luciferin sodium salt, Ultra Pure Grade
D(-)-Luciferin sodium salt
Formula: C₁₁H₇N₂NaO₃S₂
MW: 302.30 g/mol
Storage Temperature: Low Temperature Freezer (–40°C)
MDL Number: MFCD00044938
CAS Number: 103404-75-7

Order Now

Specification Test Results

Appearance Yellow powder
Water Content (KF) ≤8%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR