VWR®, SOC medium, in tubes, biotechnology grade
Supplier: VWR Chemicals
VWRCN549-6X5MLEA 1170 SGD
VWRCN549-6X5ML
VWR®, SOC medium, in tubes, biotechnology grade
Media for Microbiology Prepared Media for Microbiology
SOC medium is used in the final step of bacterial cell transformation to obtain maximum transformation efficiency of E. coli SOC.
  • Ready to use and premixed
  • Allows expression of antibiotic resistance genes
Formulation: Tryptone/casein peptone 2,0%, yeast extract 0,5%, sodium chloride 8,5 mM, potassium chloride 2,5 mM, D-glucose 20,0 mM, magnesium chloride hexahydrate 10,0 mM, magnesium sulfate heptahydrate 10,0 mM.
Order Now
SPECIFICATIONS
Environmentally Preferable