VWR® SeraFree, Cryopreservation Media
Supplier: VWR Chemicals
VWRCN676-50MLEA 628 SGD
VWRCN676-50ML
VWR® SeraFree, Cryopreservation Media
Cell Freezing Media
SeraFree cryopreservation media is ready to use freezing media for cryopreservation of adherent or suspension cultured cells. The animal-free media composition eliminates batch-to-batch variability and is optimised for cell viability and cell growth after thawing. Suitable for use in environments that prohibit the use of animal-derived products.
  • Endotoxin tested
  • Ready to use, animal-free, RPMI- or DMEM-based media
  • Optimises cell growth and cell viability after thawing
  • Reduces potential for transmission of infectious diseases