VWR® Gold-N-Gel™, RNA dye
Supplier: VWR Chemicals
VWRCN734-5MLEA 576 SGD
VWRCN734-5ML
VWR® Gold-N-Gel™, RNA dye
Stains Electrophoresis Stains
Gold-N-Gel RNA dye is compatible with standard protocols for denaturing electrophoresis of RNA fragments. Bands are immediately visible upon illumination with UV light. A standard ethidium bromide filter can be used for gel documentation.
  • Non mutagenic, fluorescent in-gel stain for immediate visualisation of RNA bands on denaturing agarose gels
  • Immediate band visualisation with UV illumination - No post-run staining or destaining
  • Easy-to-use: Simply add to melted agarose prior to casting
Delivery information: Supplied as a 200X concentrate.