VWR® Ready Ladder™ DNA Ladder, 50 bp
Supplier: VWR Chemicals
VWRCN746-100RXNEA 303 SGD
VWRCN746-100RXN
VWR® Ready Ladder™ DNA Ladder, 50 bp
Electrophoresis Reagents Molecular Weight Markers
Ladder with 10 fragments ranging from 50 to 500 bp for easy band identification. Ready to use formula pre-mixed with loading dye.