Ethyl acetate ≥99.5%

Supplier: VWR Chemicals
Danger

Synonyms: Acetic acid ethyl ester

VWRCN774-1LEA 361 SGD
VWRCN774-1L
Ethyl acetate ≥99.5%
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
UN: 1173
ADR: 3 II
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identification PASS
Purity (GC) = 99,5 %
Specific Gravity @25C 0,894 - 0,898
Water (KF) = 0,2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR