3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 10% in aqueous solution, Reagent Grade

Supplier: VWR Chemicals
Warning

VWRCN907-1LEA 1150 SGD
VWRCN907-1L
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 10% in aqueous solution, Reagent Grade
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formula: C₃₂H₅₈N₂O₇S
Storage Temperature: Refrigerator
MDL Number: MFCD00081083
CAS Number: 75621-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR