Aluminium potassium sulphate dodecahydrate 98-102% ACS

Supplier: VWR Chemicals

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

VWRCN984-500GEA 247 SGD
VWRCN984-500G
Aluminium potassium sulphate dodecahydrate 98-102% ACS
Aluminium potassium sulphate dodecahydrate
Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now

Specification Test Results

Ammonium <0.005%
Assay 98 - 102%
Chloride ≤5 ppm
Heavy Metals (as Pb) ≤0.001%
Insoluble Matter ≤0.005%
Iron ≤0.001%
Sodium ≤0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR