VWR® Electrode Holders
Supplier: VWR Chemicals
VWRCRDBT002015EA 101 SGD
VWRCRDBT002015
VWR® Electrode Holders
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Plain borosilicate tubes for insertion of a wire electrode so that the electrode is insulated from the clamp or holder and protected from mechanical damage. The tube also helps the flexible wire electrode be positioned correctly in a rigid support.
Order Now
SPECIFICATIONS
Environmentally Preferable