VWR®, Clamps, Three Pronged
Supplier: VWR Collection
VWRF08247.825EA 223 SGD
VWRF08247.825
VWR®, Clamps, Three Pronged
Clamps General Purpose Clamps
Laboral aluminium alloy.
  • Clamping fingers covered in vinyl to protect clamped items
  • Easy action knurled screw for adjusting the clamping prongs