VWR®, Clamps, Four Pronged
Supplier: VWR Collection
VWRF08297.350EA 394 SGD
VWRF08297.350
VWR®, Clamps, Four Pronged
Clamps General Purpose Clamps
Stainless steel.
  • Robust design, clamp rod diameter 12 mm, 140 mm long
  • Two easy action knurled screws for adjusting the clamping fingers independently of each other
Order Now
SPECIFICATIONS
Environmentally Preferable