VWR®, Clamps
Supplier: VWR Collection
VWRF08301.322EA 144 SGD
VWRF08301.322
VWR®, Clamps
Clamps General Purpose Clamps
Laboral aluminium alloy.
  • Round jaws covered in vinyl to protect clamped items
  • Two easy action knurled screws for adjusting each of the movable clamping jaws independently of each other