VWR®, Clamps, Three Pronged, with Flexible Rod
Supplier: VWR Collection
VWRF08303.157EA 338 SGD
VWRF08303.157
VWR®, Clamps, Three Pronged, with Flexible Rod
Clamps General Purpose Clamps
Laboral aluminium alloy.
  • Jaws covered in vinyl to protect clamped items
  • Clamp rod diameter: 8 mm; rod length: 200 mm