VWR® Stand Clamps with Flexible Rod
Supplier: VWR Collection
VWRF08309.113EA 0 SGD
VWRF08309.113
VWR® Stand Clamps with Flexible Rod
Clamps General Purpose Clamps
Laboral aluminium alloy. Clamp with flexible arm, mounting rod 12 mm Ø and rod length is 200 mm.
  • Round jaws covered in vinyl to protect clamped items
  • Convenient flexible arm has a maximum load of 500 g