VWR®, Burette Clamps
Supplier: VWR Collection
VWRF08374.287EA 117 SGD
VWRF08374.287
VWR®, Burette Clamps
Clamps General Purpose Clamps
Vinyl, black. Simple model.
  • For two burettes up to 12 mm diameter
  • Easy height adjustment
  • Can be attached to stand rods using knurled screws