VWR® Stand with Holder for Separating Funnels
Supplier: VWR Collection
VWRF08398.028EA 395 SGD
VWRF08398.028
VWR® Stand with Holder for Separating Funnels
Retort Stands
PU coated cast iron stand 200×450 mm, steel rod 500×12 mm, bosshead and holder for two funnels.
  • Height of holder can be adjusted
  • Holder measures 300×100 mm
  • Apertures for funnels measure 65 mm - centre distance 200 mm