VWR® Retort Rods, Unthreaded
Supplier: VWR Collection
VWRF08464.415EA 184 SGD
VWRF08464.415 VWRF08464.528
VWR® Retort Rods, Unthreaded
Retort Rods
Stainless steel, hollow.
  • Excellent chemical resistance
  • Durable hollow rods reducing weight in support constructions