VWR®, Curved Base Stand
Supplier: VWR Collection
VWRF08536.128EA 378 SGD
VWRF08536.128
VWR®, Curved Base Stand
Retort Stands
PU coated cast iron stand, high grade steel rod 500×12 mm.
  • Special shape of the stand ensures it is particularly solid and steady
The rod can be removed, has M8 thread, and is fixed from below with a washer and a nut.