VWR®, Large H Stands (B)
Supplier: VWR Collection
VWRF08547.217EA 835 SGD
VWRF08547.217
VWR®, Large H Stands (B)
Retort Stands
H-shaped stands with nickel-coated steel rod and 420×400 mm grey marbled cast iron baseplate.
  • H-shaped stand has adjustable baseplate
  • Designed to accommodate the upright in any one of three positions