VWR®, cell counting slides for automated cell counters
Supplier: VWR Collection
VWRI10228-0050EA 519 SGD
VWRI10228-0050
VWR®, cell counting slides for automated cell counters
Slides Counting Chambers
Slides for use with VWR® automated cell counters.
  • Disposable slides with throughput of 2 samples/slide
  • For research use only