VWR®, Autoclavable Cleanroom Goggles
Supplier: VWR Collection
VWRI111-0614EA 0 SGD
VWRI111-0614
VWR®, Autoclavable Cleanroom Goggles
Goggles
Lightweight cleanroom goggles.
  • Soft, thermoplastic body with toughened polycarbonate lens
  • Strap with polypropylene strap hooks
  • Indirect base ventilation
  • Autoclavable at 121 °C
Suitable for ISO Class 4 environments.
Certifications: EN 166:2001; Optical Class 1.
Packaging: Packed in a NEBB Certified ISO Class 4 (Class 10) cleanroom. One piece per PE bag.