VWR® Lisbon, Visitor Spectacles
Supplier: VWR Collection
VWRI111-0930EA 7.1 SGD
VWRI111-0930
VWR® Lisbon, Visitor Spectacles
Safety Glasses
Visitor glasses with wrap-around, clear polycarbonate lens and indirect ventilation.
  • Anti-scratch/anti-fog coating
  • Integrated brow guard
  • Vented side shields
Weight: 46 g
Weight: 46 g
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens).