VWR® Montreal, Safety Goggles
Supplier: VWR Collection
VWRI111-1839EA 12.9 SGD
VWRI111-1839
VWR® Montreal, Safety Goggles
Goggles
Ergonomically designed wide vision goggles with co-injected frame provides 180° panoramic view. PC lens, PP (hard part) and AES Santoprene (soft part) frame, polyester-rubber headband.
  • Indirect ventilation to prevent fogging
  • Withstands extreme temperatures
  • Designed to give optimum fit to all facial contours
  • Easy four-step lens replacement
Certifications: Compliant with: EN 166:2001. Frame: EN 166 3 9 BT CE. Lens: VWR® 1 BT 9.