VWR® Toronto, Safety Goggles
Supplier: VWR Collection
VWRI111-1840EA 11.8 SGD
VWRI111-1840
VWR® Toronto, Safety Goggles
Goggles
Wrap-around modern style provides excellent panoramic vision, high level of comfort and maximum protection.
  • PC lens, PVC frame, polyester/rubber headband
  • Fitted with indirect ventilation
  • Opalescent frame prevents light reflection
  • Designed to give optimum fit to all facial contours
  • Comfortably fits over most spectacles
Certifications: Compliant with: EN 166:2001. Frame: EN166 3 4 9 BT CE. Lens: VWR® 1 BT 9.